Tungsten(VI) fluoride

Synonyms: Tungsten hexafluoride

399108-225G 399108-450G
220064-119EA 2661.54 USD
220064-119 220064-120
Tungsten(VI) fluoride
Tungsten(VI) fluoride
Formula: F₆W
MW: 297.83 g/mol
Boiling Pt: 17.5 °C (1013 hPa)
Melting Pt: 3 °C
Density: 12.9 g/cm³ (20 °C)
CAS Number: 7783-82-6
EINECS: 232-029-1
Merck Index: 12,09946

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR