3-Hexylthiophene

399051-1G 399051-5G
220064-116EA 54.28 USD
220064-116 220064-117
3-Hexylthiophene
3-Hexylthiophene
Formula: C₁₀H₁₆S
MW: 168.3 g/mol
Boiling Pt: 60…61 °C (0.8 mmHg)
Density: 0.9392 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00143181
CAS Number: 1693-86-3
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR