Osmium(III) chloride

Synonyms: Osmium trichloride

398594-100MG 398594-500MG
220064-080EA 144.15 USD
220064-080 220064-081
Osmium(III) chloride
Osmium(III) chloride
Formula: Cl₃Os
MW: 296.59 g/mol
Melting Pt: 500 °C
Density: 5.3 g/cm³ (20 °C)
MDL Number: MFCD00011148
CAS Number: 13444-93-4
EINECS: 236-587-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR