Gallic acid monohydrate ≥98.0% ACS

Warning

Synonyms: 3,4,5-Trihydroxybenzoic acid monohydrate, 3,4,5-Trihydroxybenzoic acid hydrate

398225-100G 398225-500G
220064-059EA 73.62 USD
220064-059 220064-060
Gallic acid monohydrate ≥98.0% ACS
Gallic acid monohydrate
Formula: C₇H₆O₅·1H₂O
MW: 188.14 g/mol
Melting Pt: ∼250 °C
Flash Pt: 250 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00149098
CAS Number: 5995-86-8
EINECS: 205-749-9

Order Now

Specification Test Results

Assay 98.0%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR