Xylenol orange tetrasodium salt ACS

Synonyms: Xylenol orange water soluble, 2,2',2'',2'''-((((1,1-Dioxido-3H-benzo[c][1,2]oxathiole-3,3-diyl)bis(6-hydroxy-5-methyl-3,1-phenylene))bis(methylene))bis(azanetriyl))tetraacetic acid, sodium salt (1:4)

398187-25G 398187-5G 398187-1G
220063-989EA 721 USD
220063-989 220063-990 220063-988
Xylenol orange tetrasodium salt ACS
Xylenol orange tetrasodium salt
Formula: C₃₁H₂₈N₂Na₄O₁₃S
MW: 760.59 g/mol
Melting Pt: ∼210 °C
Storage Temperature: Ambient
MDL Number: MFCD00044293
CAS Number: 3618-43-7
EINECS: 222-805-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=132190c5346f09a24d8f812d8fde344fb5a92ad5&vid=e38e97eec95747d23a2c3c9d900a009b87f38e99&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR