Lithium sulfate monohydrate ≥99% ACS

Warning

398152-100G 398152-500G
220063-517EA 54.15 USD
220063-517 220063-518
Lithium sulfate monohydrate ≥99% ACS
Lithium sulphate monohydrate
Formula: Li₂SO₄·H₂O
MW: 127.96 g/mol
Melting Pt: 120 °C
Density: 2.05 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149766
CAS Number: 10102-25-7
EINECS: 233-820-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR