Sodium dichromate dihydrate ≥99.5% ACS

Danger

398063-500G 398063-100G
220063-943EA 160.62 USD
220063-943 220063-941
Sodium dichromate dihydrate ≥99.5% ACS
Sodium dichromate dihydrate
Formula: Na₂Cr₂O₇·2H₂O
Melting Pt: 356.7 °C
Density: 2.35 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149166
CAS Number: 7789-12-0
EINECS: 234-190-3
UN: 3087
ADR: 5.1,II
Merck Index: 12,08754

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR