2,2,2-Trifluoroethanol-[D₃]

Synonyms: 2,2,2-Trifluoroethanol-[²H₃], TFE-D3

396532-1G 396532-5G
220063-871EA 49.23 USD
220063-871 220063-872
2,2,2-Trifluoroethanol-[D₃]
2,2,2-Trifluoroethanol-[D₃]
Formula: F₃CCD₂OD
MW: 103.02 g/mol
Boiling Pt: 75 °C (1013 hPa)
Melting Pt: –45 °C
Density: 1.45 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00037682
CAS Number: 77253-67-9
EINECS: 278-649-6
UN: 1992
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR