1-Chloro-4-iodobutane ≥98%

396222-25ML 396222-5ML
220063-841EA 396.31 USD
220063-841 220063-842
1-Chloro-4-iodobutane ≥98%
1-Chloro-4-iodobutane
Formula: C₄H₈ClI
MW: 218.46 g/mol
MDL Number: MFCD00039415
CAS Number: 10297-05-9
EINECS: 233-669-4

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR