Strontium titanate ≥99%

396141-500G 396141-100G
220063-833EA 238.46 USD
220063-833 220063-832
Strontium titanate ≥99%
Strontium titanate
Formula: O₃SrTi
MW: 183.5 g/mol
Melting Pt: 2080 °C
Density: 5.11 g/cm³ (20 °C)
MDL Number: MFCD00049554
CAS Number: 12060-59-2
EINECS: 235-044-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR