Tris(4-methoxyphenyl)phosphine ≥95%

Synonyms: Tris(p-methoxyphenyl)phosphine

395102-5G 395102-1G
220063-780EA 109.38 USD
220063-780 220063-779
Tris(4-methoxyphenyl)phosphine ≥95%
Tris(4-methoxyphenyl)phosphine
Formula: C₂₁H₂₁O₃P
MW: 352.37 g/mol
Boiling Pt: 446 °C (1013 hPa)
Melting Pt: 135 °C
MDL Number: MFCD00014896
CAS Number: 855-38-9
EINECS: 212-723-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR