2-(2,4-Dichlorophenyl)ethanol ≥97%

Synonyms: 2,4-Dichlorophenethyl alcohol

394556-5G 394556-1G
220062-982EA 110.77 USD
220062-982 220062-981
2-(2,4-Dichlorophenyl)ethanol ≥97%
2-(2,4-Dichlorophenyl)ethanol
Formula: C₈H₈Cl₂O
MW: 191.06 g/mol
MDL Number: MFCD00192010
CAS Number: 81156-68-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR