Xenon difluoride ≥99.99%

Danger

394505-1G 394505-5G
220063-595EA 294.23 USD
220063-595 220063-702
Xenon difluoride ≥99.99%
Xenon difluoride
Formula: F₂Xe
Boiling Pt: 114 °C (1013 hPa)
Density: 4.32 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00040538
CAS Number: 13709-36-9
EINECS: 237-251-2
UN: 3087
ADR: 5.1,II
Merck Index: 14,10072

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR