4-Bromo-2,6-di-tert-butylphenol ≥98%

Synonyms: 1-Bromo-3,5-di(dimethylethyl)-4-hydroxybenzene

394378-25G 394378-100G
220061-171EA 154 USD
220061-171 220061-170
4-Bromo-2,6-di-tert-butylphenol ≥98%
4-Bromo-2,6-di-tert-butylphenol
Formula: C₁₄H₂₁BrO
MW: 285.22 g/mol
Boiling Pt: 104…107 °C (1 mmHg)
Melting Pt: 83 °C
MDL Number: MFCD00051598
CAS Number: 1139-52-2
EINECS: 214-521-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR