EDC (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide)

Synonyms: EDAC, WSC, EDC, N'-(Ethylcarbonimidoyl)-N,N-dimethylpropane-1,3-diamine, 1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide, N1-((Ethylimino)methylene)-N3,N3-dimethylpropane-1,3-diamine

39391-10ML 39391-50ML
220063-564EA 191.69 USD
220063-564 220063-565
EDC (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide)
EDC (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide)
Formula: C₈H₁₇N₃
MW: 155.24 g/mol
Boiling Pt: 43 - 45 °C (0.3 -rr)
Melting Pt: 114 - 115 °C
Density: 0.885 g/cm³ (20 °C)
MDL Number: MFCD00044916
CAS Number: 1892-57-5
EINECS: 217-579-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR