Gallium(III) acetylacetonate

Synonyms: Tris(2,4-pentanedionato-O,O')gallium (III), Gallium (III) 2,4-pentanedionate, Gallium (III) tris(2,4-pentanedionato-O,O'), Gallium(III) Acetylacetonate, Acetylacetone Gallium(III) salt, Tris(2,4-pentanedionato)gallium(III), Ga(acac)3

393541-5G 393541-25G
220063-531EA 256.92 USD
220063-531 220063-530
Gallium(III) acetylacetonate
Gallium(III) acetylacetonate
Formula: C₁₅H₂₁GaO₆
MW: 367.05 g/mol
Melting Pt: 196…198 °C
Density: 1.42 g/cm³ (20 °C)
MDL Number: MFCD00013492
CAS Number: 14405-43-7
EINECS: 238-377-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR