Tris(dimethylamino)phosphine ≥97%

Synonyms: Tris(dimethylamino)phosphine, Hexamethyl phosphorous triamide, Hexamethyltriaminophosphine, HMPT

393290-25ML 393290-5ML
220063-466EA 191.77 USD
220063-466 220063-467
Tris(dimethylamino)phosphine ≥97%
Tris(dimethylamino)phosphine
Formula: C₆H₁₈N₃P
MW: 163.2 g/mol
Boiling Pt: 150 °C (1013 hPa)
Melting Pt: –44 °C
Density: 0.8962 g/cm³ (20 °C)
MDL Number: MFCD00008301
CAS Number: 1608-26-0
EINECS: 216-534-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR