5-Iodo-m-xylene ≥99%

Warning

Synonyms: 1-Iodo-3,5-dimethylbenzene, 3,5-Dimethyl-1-iodobenzene, 1,3-Dimethyl-5-iodobenzene, 3,5-Dimethyliodobenzene

392510-25ML 392510-5ML
220063-412EA 133.54 USD
220063-412 220063-413
5-Iodo-m-xylene ≥99%
5-Iodo-m-xylene
Formula: C₈H₉I
MW: 232.06 g/mol
Boiling Pt: 234…235 °C (1013 hPa)
Density: 1.61 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00060659
CAS Number: 22445-41-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR