ω,4-Dichloroacetophenone ≥98%

Synonyms: 2-Chloro-1-(4-chlorophenyl)ethanone, α,4-Dichloroacetophenone, 2,4'-Dichloroacetophenone

392006-100G 392006-25G
220061-164EA 314.46 USD
220061-164 220061-165
ω,4-Dichloroacetophenone ≥98%
ω,4-Dichloroacetophenone
Formula: C₈H₆Cl₂O
MW: 189.04 g/mol
Boiling Pt: 270 °C (1013 hPa)
Melting Pt: 98 °C
MDL Number: MFCD00018926
CAS Number: 937-20-2
EINECS: 213-323-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR