tert-Butylamine ≥99.5%

Danger

Synonyms: 2-Aminoisobutane

391433-100ML 391433-1L
220060-563EA 102.31 USD
220060-563 220060-564
tert-Butylamine ≥99.5%
tert-Butylamine
Formula: C₄H₁₁N
MW: 73.14 g/mol
Boiling Pt: 44.4 °C (1013 hPa)
Melting Pt: –72 °C
Density: 0.6965 g/cm³ (20 °C)
Flash Pt: –38 °C
Storage Temperature: Ambient
MDL Number: MFCD00008050
CAS Number: 75-64-9
EINECS: 200-888-1
UN: 2734
ADR: 8,I
Merck Index: 13,01544

Order Now

Specification Test Results

Assay 99.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR