N,N-Dimethylethylendiamine ≥98.0% (by GC)

Danger

Synonyms: N,N-Dimethylethylenediamine, asym. Dimethylethylendiamine, unsym-Dimethylethylenediamine, 2-(Dimethylamino)ethylamine, 1-Amino-2-dimethylaminoethane, 2-Dimethylaminoethylamine, 2-Aminoethyldimethylamine

39030-500ML 39030-100ML
220063-325EA 220.77 USD
220063-325 220063-324
N,N-Dimethylethylendiamine ≥98.0% (by GC)
N,N-Dimethylethylendiamine
Formula: C₄H₁₂N₂
MW: 88.15 g/mol
Boiling Pt: ~ 106 °C (1013 hPa)
Melting Pt: -70 °C
Density: 0.804 g/cm³ (20 °C)
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00008175
CAS Number: 108-00-9
EINECS: 203-541-2

Order Now

Specification Test Results

Assay 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR