3,5-Dimethylphenyl isocyanate ≥99%

Synonyms: 3,5-Xylyl isocyanate, 3,5-Dimethylphenylisocyanate

390100-25G 390100-5G
220063-308EA 137.85 USD
220063-308 220063-309
3,5-Dimethylphenyl isocyanate ≥99%
3,5-Dimethylphenyl isocyanate
Formula: C₉H₉NO
MW: 147.18 g/mol
Boiling Pt: 205 °C (1013 hPa)
Density: 1.04 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00013868
CAS Number: 54132-75-1
EINECS: 258-987-0
UN: 2206
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR