δ-Valerolactone, Technical Grade

Synonyms: Pentan-5-olide, Tetrahydro-2H-pyran-2-one, 5-Hydroxypentanoic acid lactone, δ-Valerolactone

389579-100ML 389579-25ML
220063-276EA 170.15 USD
220063-276 220063-277
δ-Valerolactone, Technical Grade
δ-Valerolactone
Formula: C₅H₈O₂
MW: 100.12 g/mol
Boiling Pt: 228…230 °C (1013 hPa)
Melting Pt: –10 °C
Density: 1.0988 g/cm³ (20 °C)
MDL Number: MFCD00006645
CAS Number: 542-28-9
EINECS: 208-807-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR