ω-Bromo-4-fluoroacetophenone ≥97%

Synonyms: Bromomethyl p-fluorophenyl ketone, α-Bromo-4-fluoroacetophenone, 2-Bromo-1-(4-fluorophenyl)ethan-1-one, 2-Bromo-4'-fluoroacetophenone, 4-Fluorophenacyl bromide, 2-Bromo-1-(4-fluorophenyl)ethanone

389390-5G 389390-25G
220063-259EA 63 USD
220063-259 220063-258
ω-Bromo-4-fluoroacetophenone ≥97%
ω-Bromo-4-fluoroacetophenone
Formula: C₈H₆BrFO
MW: 217.04 g/mol
Boiling Pt: 82 °C (0.4 mmHg)
Melting Pt: 49 °C
MDL Number: MFCD00040830
CAS Number: 403-29-2
EINECS: 206-955-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR