1-Butyl-3-methylimidazolium chloride, Sigma-Aldrich®

Synonyms: 3-Butyl-1-methyl-1H-imidazol-3-ium chloride

38899-100G-F 38899-1KG-F
220062-166EA 513.23 USD
220062-166 220062-167
1-Butyl-3-methylimidazolium chloride, Sigma-Aldrich®
1-Butyl-3-methylimidazolium chloride
Formula: C₈H₁₅ClN₂
MW: 174.67 g/mol
Boiling Pt: ∼200 °C (1013 hPa)
Melting Pt: ∼60 °C
Density: ~ 1 g/cm³ (20 °C)
Flash Pt: >110 °C
MDL Number: MFCD03095425
CAS Number: 79917-90-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR