Methyl-2-bromobenzoate ≥98%

Synonyms: 2-Bromobenzoic acid methyl ester, Methyl 2-bromobenzoate

387851-25ML 387851-5ML
220063-194EA 61.92 USD
220063-194 220063-195
Methyl-2-bromobenzoate ≥98%
Methyl-2-bromobenzoate
Formula: C₈H₇BrO₂
MW: 215.05 g/mol
Boiling Pt: 253 °C (1013 hPa)
Density: 1.51 g/cm³ (20 °C)
MDL Number: MFCD00016328
CAS Number: 610-94-6
EINECS: 210-241-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR