N-Ethyldiisopropylamine ≥99.5%

Danger

387649-100ML 387649-1L
220062-959EA 66.77 USD
220062-959 220062-960
N-Ethyldiisopropylamine ≥99.5%
N-Ethyldiisopropylamine
Formula: C₈H₁₉N
Boiling Pt: 127 °C (1013 hPa)
Melting Pt: –50 °C
Density: 0.76 g/cm³ (20 °C)
Flash Pt: 6 °C
Storage Temperature: Ambient
MDL Number: MFCD00008868
CAS Number: 7087-68-5
EINECS: 230-392-0
UN: 2733
ADR: 3,II

Order Now

Specification Test Results

Assay 99.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR