4-(Trifluoromethoxy)benzenesulfonyl chloride ≥98%

Synonyms: 4-(Trifluoromethoxy)benzene-1-sulphonyl chloride

386545-5G 386545-1G
220063-152EA 46.2 USD
220063-152 220063-151
4-(Trifluoromethoxy)benzenesulfonyl chloride ≥98%
4-(Trifluoromethoxy)benzenesulphonyl chloride
Formula: C₇H₄ClF₃O₃S
MW: 260.62 g/mol
Boiling Pt: 120…122 °C (15 mmHg)
Density: 1.536 g/cm³ (20 °C)
MDL Number: MFCD00042408
CAS Number: 94108-56-2
EINECS: 302-397-9
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR