3-Methoxy-1-propanol ≥98.0% (by GC)

Synonyms: Trimethylene glycol monomethyl ether

38457-10ML-F 38457-50ML-F
220061-943EA 103.17 USD
220061-943 220061-944
3-Methoxy-1-propanol ≥98.0% (by GC)
3-Methoxy-1-propanol
Formula: C₄H₁₀O₂
MW: 90.12 g/mol
MDL Number: MFCD00039550
CAS Number: 1589-49-7
EINECS: 433-230-9
UN: 1987
ADR: 3,III

Order Now

Specification Test Results

Assay 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR