4-Octylphenol ≥99%

384445-1G 384445-5G
220061-939EA 66.12 USD
220061-939 220061-940
4-Octylphenol ≥99%
4-Octylphenol
Formula: C₁₄H₂₂O
MW: 206.33 g/mol
Boiling Pt: 315 °C (1013 hPa)
Melting Pt: 38 - 39 °C
Density: 0.961 g/cm³ (20 °C)
MDL Number: MFCD00036134
CAS Number: 1806-26-4
EINECS: 217-302-5
UN: 2430
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR