4-tert-Butylbenzaldehyde ≥97%

Synonyms: 4-(tert-Butyl)benzaldehyde

384038-25ML 384038-5ML
220063-039EA 54.6 USD
220063-039 220063-040
4-tert-Butylbenzaldehyde ≥97%
4-tert-Butylbenzaldehyde
Formula: C₁₁H₁₄O
MW: 162.23 g/mol
Boiling Pt: 58 - 60 °C (0.2 -rr)
Density: 0.971 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00035742
CAS Number: 939-97-9
EINECS: 213-367-9
UN: 3082
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR