5'-Chloro-2'-hydroxyacetophenone ≥99%

Synonyms: 1-(5-Chloro-2-hydroxyphenyl)ethan-1-one, 1-(5-Chloro-2-hydroxyphenyl)ethanone

383384-25G 383384-5G
220063-011EA 48.46 USD
220063-011 220063-012
5'-Chloro-2'-hydroxyacetophenone ≥99%
5'-Chloro-2'-hydroxyacetophenone
Formula: C₈H₇ClO₂
MW: 170.6 g/mol
MDL Number: MFCD00067788
CAS Number: 1450-74-4
EINECS: 215-916-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR