Zinc acetate ≥99.99%

Warning

383317-25G 383317-100G
220060-082EA 72 USD
220060-082 220060-081
Zinc acetate ≥99.99%
Zinc acetate
Formula: C₄H₆O₄Zn
MW: 183.48 g/mol
Boiling Pt: 908 °C (1013 hPa)
Density: 1.735 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00012454
CAS Number: 557-34-6
EINECS: 209-170-2
UN: 3077
ADR: 9,III
Merck Index: 12,10256

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR