Sodium iodide ≥99.5% ACS

Warning

Synonyms: Hydriodic acid sodium salt, Sodium monoiodide, Ioduril

383112-2.5KG 383112-500G 383112-100G
220062-994EA 627.69 USD
220062-994 220062-995 220062-993
Sodium iodide ≥99.5% ACS
Sodium iodide
Formula: NaI
MW: 149.89 g/mol
Boiling Pt: 1300 °C (1013 hPa)
Melting Pt: 662 °C
Density: 3.67 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003532
CAS Number: 7681-82-5
EINECS: 231-679-3
UN: 3077
ADR: 9,III
Merck Index: 13,08705

Order Now

Specification Test Results

Assay 99.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR