Ammonium cerium (IV) sulfate dihydrate ACS

Synonyms: Tetraammonium cerium tetrakis(sulphate) dihydrate, Cerium (IV) ammonium sulphate dihydrate, Tetraammonium cerium(IV) sulphate dihydrate, Ammonium cerium(IV) sulphate hydrate, Ammonium ceric sulphate dihydrate

383090-500G 383090-100G
220062-941EA 304.62 USD
220062-941 220062-940
Ammonium cerium (IV) sulfate dihydrate ACS
Ammonium cerium(IV) sulphate dihydrate
Formula: (NH₄)₂Ce(SO₄)₄·2H₂O
MW: 632.55 g/mol
Melting Pt: 130 °C
MDL Number: MFCD00149417
CAS Number: 10378-47-9
EINECS: 231-567-4

Order Now

Specification Test Results

Assay 94

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR