Lead(II) perchlorate trihydrate ≥98% ACS

Synonyms: Lead diperchlorate trihydrate

383066-500G 383066-100G
220062-932EA 143.38 USD
220062-932 220062-931
Lead(II) perchlorate trihydrate ≥98% ACS
Lead(II) perchlorate trihydrate
Formula: Cl₂O₈Pb·3H₂O
MW: 460.15 g/mol
Melting Pt: 280 °C
Density: 4.82 to 4.86 g/cm³ (25 °C)
MDL Number: MFCD00149817
CAS Number: 13453-62-8
EINECS: 237-125-7
UN: 1470
ADR: 5.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR