1,3-Difluoro-2-nitrobenzene ≥98%

Danger

Synonyms: 2,6-Difluoronitrobenzene

382957-1G 382957-5G
220062-925EA 26.92 USD
220062-925 220062-926
1,3-Difluoro-2-nitrobenzene ≥98%
1,3-Difluoro-2-nitrobenzene
Formula: C₆H₃F₂NO₂
MW: 159.09 g/mol
MDL Number: MFCD00192035
CAS Number: 19064-24-5
EINECS: 242-793-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR