4-(Hydroxymethyl)benzoic acid ≥99%

Synonyms: 4-Carboxybenzylalcohol, HMBA Linker, HMBA

382639-10G 382639-1G
220062-899EA 144.31 USD
220062-899 220062-900
4-(Hydroxymethyl)benzoic acid ≥99%
4-(Hydroxymethyl)benzoic acid
Formula: C₈H₈O₃
MW: 152.15 g/mol
MDL Number: MFCD00017598
CAS Number: 3006-96-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR