Zirconyl nitrate hydrate

Danger

Synonyms: Zirconium(IV) oxynitrate hydrate, Zirconium(IV) dinitrate oxide hydrate, Zirconium dinitrate oxide hydrate

380679-50G 380679-10G
220062-735EA 627.69 USD
220062-735 220062-340
Zirconyl nitrate hydrate
Zirconyl nitrate hydrate
Formula: N₂O₇Zr·H₂O
MW: 249.25 g/mol
Density: 2.04 g/cm³ (20 °C)
MDL Number: MFCD00149896
CAS Number: 14985-18-3
EINECS: 237-529-3
UN: 1477
ADR: 5.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR