2-Amino-5-methylthiazole

Synonyms: 5-Methylthiazol-2-ylamine

380563-25G 380563-5G
220062-325EA 74.74 USD
220062-325 220062-326
2-Amino-5-methylthiazole
2-Amino-5-methylthiazole
Formula: C₄H₆N₂S
MW: 114.17 g/mol
Boiling Pt: 222 °C (1013 hPa)
Melting Pt: 96 °C
MDL Number: MFCD00078317
CAS Number: 7305-71-7
EINECS: 423-800-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR