4,4-Dimethoxybutanone ≥90%, Technical Grade

Synonyms: 4,4-Dimethoxybutan-2-one, 3-Oxobutyraldehyde dimethyl acetal, Acetoacetaldehyde-1,1-Dimethyl Acetal, Acetylacetaldehyde dimethyl acetal, 4,4-Dimethoxy-2-butanone

380067-100ML 380067-500ML
220062-701EA 94.45 USD
220062-701 220062-702
4,4-Dimethoxybutanone ≥90%, Technical Grade
4,4-Dimethoxybutanone
Formula: C₆H₁₂O₃
MW: 132.16 g/mol
Boiling Pt: 178 °C (1013 hPa)
Melting Pt: –82 °C
Density: 0.9451 g/cm³ (20 °C)
Flash Pt: 58 °C
MDL Number: MFCD00008789
CAS Number: 5436-21-5
EINECS: 226-605-1
UN: 1224
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR