Iron(II) chloride tetrahydrate

Danger

Synonyms: Ferrous chloride tetrahydrate, Iron dichloride tetrahydrate

380024-5G 380024-25G
220062-696EA 87.08 USD
220062-696 220062-695
Iron(II) chloride tetrahydrate
Iron(II) chloride tetrahydrate
Formula: FeCl₂·4H₂O
MW: 198.81 g/mol
Melting Pt: 677 °C
Density: 1.926 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149709
CAS Number: 13478-10-9
EINECS: 231-843-4
UN: 3260
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR