Diammonium hydrogen orthophosphate

379980-25G 379980-100G
220062-686EA 161.54 USD
220062-686 220062-685
Diammonium hydrogen orthophosphate
di-Ammonium hydrogen orthophosphate
Formula: (NH₄)₂HPO₄
Melting Pt: 185 °C
Density: 1.619 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010891
CAS Number: 7783-28-0
EINECS: 231-987-8
REACH: 01-2119490974-22
Merck Index: 13,00546

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR