Nickel acetate tetrahydrate ≥99.998%

Danger

Synonyms: Nickel diacetate tetrahydrate, Nickel (II) acetate tetrahydrate, Nickel di(acetate) tetrahydrate, Acetic acid nickel (II) salt tetrahydrate

379883-10G 379883-50G
220062-672EA 146.46 USD
220062-672 220062-673
Nickel acetate tetrahydrate ≥99.998%
Nickel acetate tetrahydrate
Formula: C₄H₆NiO₄·4H₂O
MW: 248.84 g/mol
Boiling Pt: 16 °C (1013 hPa)
Density: 1.798 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00066973
CAS Number: 6018-89-9
EINECS: 206-761-7
Merck Index: 12,06583

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR