Palladium(II) acetate ≥99.98%

Synonyms: Palladium diacetate, Palladium (II) acetate

379875-5G 379875-1G
220062-671EA 1120 USD
220062-671 220062-670
Palladium(II) acetate ≥99.98%
Palladium(II) acetate
Formula: C₄H₆O₄Pd
MW: 224.51 g/mol
Melting Pt: 205 °C
MDL Number: MFCD00012453
CAS Number: 3375-31-3
EINECS: 222-164-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR