di-Ammonium oxalate monohydrate ≥99.99%

Warning

Synonyms: Ammonium oxalate monohydrate, Ammonium oxalate-1-hydrate, Oxalic acid diammonium salt monohydrate, Oxalic acid ammonium salt monohydrate, Ammonium ethanedioate monohydrate, Ammonium Oxalate, Monohydrate

379743-25G 379743-5G
220062-650EA 303.08 USD
220062-650 220062-651
di-Ammonium oxalate monohydrate ≥99.99%
di-Ammonium oxalate monohydrate
Formula: (NH₄)₂C₂O₄·H₂O
MW: 142.11 g/mol
Boiling Pt: 70 °C (1013 hPa)
Melting Pt: 70 °C
Density: ~ 1.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149694
CAS Number: 6009-70-7
EINECS: 214-202-3
UN: 3288
ADR: 6.1,III
Merck Index: 13,00541

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR