HMDS (1,1,1,3,3,3-hexamethyldisilazane), ReagentPlus®

Synonyms: Hexamethyldisilazane

379212-25ML 379212-100ML
220062-123EA 73.08 USD
220062-123 220062-122
HMDS (1,1,1,3,3,3-hexamethyldisilazane), ReagentPlus®
HMDS (1,1,1,3,3,3-hexamethyldisilazane)
Formula: (CH₃)₃SiNHSi(CH₃)₃
MW: 161.39 g/mol
MDL Number: MFCD00008259
CAS Number: 999-97-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ce35a38a4179292c4ff37b2f3093018738545e6e&vid=1693ad54a45ca3ae50a06056893e8438d1002549&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR