2,3-Dihydroxynaphthalene

Synonyms: Naphthalene-2,3-diol, 2,3-Naphthalenediol

37760-250G-F 37760-50G-F
220062-470EA 235.85 USD
220062-470 220062-471
2,3-Dihydroxynaphthalene
2,3-Dihydroxynaphthalene
Formula: C₁₀H₈O₂
MW: 160.17 g/mol
Boiling Pt: 354 °C (1013 hPa)
Melting Pt: 151…152 °C
Density: 1.12 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004073
CAS Number: 92-44-4
EINECS: 202-156-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR