3,4-Dimethoxy-3-cyclobutene-1,2-dione ≥99%

Synonyms: 1,2-Dimethoxy-1-cyclobutene-3,4-dione, Dimethyl squarate, Squaric acid dimethylester

377406-1G 377406-5G
220062-451EA 28.43 USD
220062-451 220062-452
3,4-Dimethoxy-3-cyclobutene-1,2-dione ≥99%
3,4-Dimethoxy-3-cyclobutene-1,2-dione
Formula: C₆H₆O₄
MW: 142.11 g/mol
Boiling Pt: 247 °C (1013 hPa)
Melting Pt: 55 °C
MDL Number: MFCD00101316
CAS Number: 5222-73-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR