2,6-Diisopropylphenyl isocyanate ≥98%

Synonyms: 2,6-Diisopropylphenylisocyanate

376930-5G 376930-1G
220062-284EA 88.52 USD
220062-284 220062-283
2,6-Diisopropylphenyl isocyanate ≥98%
2,6-Diisopropylphenyl isocyanate
Formula: C₁₃H₁₇NO
MW: 203.28 g/mol
Boiling Pt: 117 °C (1013 hPa)
Density: 1.2485 g/cm³ (20 °C)
MDL Number: MFCD00008882
CAS Number: 28178-42-9
EINECS: 248-885-4
UN: 3382
ADR: 6.1,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR